Merck
  • Sustainable Magnetic Materials (from Chitosan and Municipal Biowaste) for the Removal of Diclofenac from Water.
Sustainable Magnetic Materials (from Chitosan and Municipal Biowaste) for the Removal of Diclofenac from Water.
Nanomaterials (Basel, Switzerland) (2019-08-02)
Roberto Nisticò, Alessandra Bianco Prevot, Giuliana Magnacca, Lorenzo Canone, Sara García-Ballesteros, Antonio Arques
ABSTRACT
The photodegradation of an aqueous solution of diclofenac (DCF) has been attempted in the presence of hydrogen peroxide and organic/inorganic hybrid magnetic materials under simulated and real solar light. The hybrid magnetic materials have been prepared via coprecipitation synthesis starting from iron(II) and iron(III) inorganic salts in the presence of bioderived organic products (i.e., chitosan or bio-based substances isolated from commercially available composted urban biowastes) acting as stabilizers of the iron-containing phase. In addition to the as prepared hybrid materials, the corresponding materials obtained after a pyrolytic step at low temperature (550 °C) have been tested. The obtained results evidenced the capability of the materials to activate hydrogen peroxide at mild pH promoting DCF (photo) degradation. All the materials feature also as adsorbents since a decrease of DCF is observed also when working in the dark and in the absence of hydrogen peroxide.
Product Description
Supelco
Acetonitrile, for UHPLC-MS LiChrosolv®
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, for UHPLC, suitable for mass spectrometry (MS)
Pricing and availability is not currently available.