Merck
  • Acyl chain asymmetry and polyunsaturation of brain phospholipids facilitate membrane vesiculation without leakage.
Acyl chain asymmetry and polyunsaturation of brain phospholipids facilitate membrane vesiculation without leakage.
eLife (2018-03-16)
Marco M Manni, Marion L Tiberti, Sophie Pagnotta, Hélène Barelli, Romain Gautier, Bruno Antonny
ABSTRACT
Phospholipid membranes form cellular barriers but need to be flexible enough to divide by fission. Phospholipids generally contain a saturated fatty acid (FA) at position sn1 whereas the sn2-FA is saturated, monounsaturated or polyunsaturated. Our understanding of the impact of phospholipid unsaturation on membrane flexibility and fission is fragmentary. Here, we provide a comprehensive view of the effects of the FA profile of phospholipids on membrane vesiculation by dynamin and endophilin. Coupled to simulations, this analysis indicates that: (i) phospholipids with two polyunsaturated FAs make membranes prone to vesiculation but highly permeable; (ii) asymmetric sn1-saturated-sn2-polyunsaturated phospholipids provide a tradeoff between efficient membrane vesiculation and low membrane permeability; (iii) When incorporated into phospholipids, docosahexaenoic acid (DHA; omega-3) makes membranes more deformable than arachidonic acid (omega-6). These results suggest an explanation for the abundance of sn1-saturated-sn2-DHA phospholipids in synaptic membranes and for the importance of the omega-6/omega-3 ratio on neuronal functions.
Product Description
Avanti
20:4 (Cis) PC, 1,2-diarachidonoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
22:6 (Cis) PC, 1,2-didocosahexaenoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
16:0-22:6 PS, 1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform
Avanti
22:6 PS, 1,2-didocosahexaenoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform
Avanti
18:0-20:4 PS, 1-stearoyl-2-arachidonoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform
Avanti
18:0-18:1 PS, 1-stearoyl-2-oleoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform
Avanti
18:0-18:1 PE, 1-stearoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine, powder
Avanti
16:0-20:4 PS, 1-palmitoyl-2-arachidonoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform
Avanti
18:0-22:6 PS, 1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform
Avanti
18:0-18:2 PS, 1-stearoyl-2-linoleoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform
Avanti
18:0-18:2 PS, 1-stearoyl-2-linoleoyl-sn-glycero-3-phospho-L-serine (sodium salt), powder