Merck
  • The role of Mediator and Little Elongation Complex in transcription termination.
The role of Mediator and Little Elongation Complex in transcription termination.
Nature communications (2020-02-28)
Hidehisa Takahashi, Amol Ranjan, Shiyuan Chen, Hidefumi Suzuki, Mio Shibata, Tomonori Hirose, Hiroko Hirose, Kazunori Sasaki, Ryota Abe, Kai Chen, Yanfeng He, Ying Zhang, Ichigaku Takigawa, Tadasuke Tsukiyama, Masashi Watanabe, Satoshi Fujii, Midori Iida, Junichi Yamamoto, Yuki Yamaguchi, Yutaka Suzuki, Masaki Matsumoto, Keiichi I Nakayama, Michael P Washburn, Anita Saraf, Laurence Florens, Shigeo Sato, Chieri Tomomori-Sato, Ronald C Conaway, Joan W Conaway, Shigetsugu Hatakeyama
ABSTRACT
Mediator is a coregulatory complex that regulates transcription of Pol II-dependent genes. Previously, we showed that human Mediator subunit MED26 plays a role in the recruitment of Super Elongation Complex (SEC) or Little Elongation Complex (LEC) to regulate the expression of certain genes. MED26 plays a role in recruiting SEC to protein-coding genes including c-myc and LEC to small nuclear RNA (snRNA) genes. However, how MED26 engages SEC or LEC to regulate distinct genes is unclear. Here, we provide evidence that MED26 recruits LEC to modulate transcription termination of non-polyadenylated transcripts including snRNAs and mRNAs encoding replication-dependent histone (RDH) at Cajal bodies. Our findings indicate that LEC recruited by MED26 promotes efficient transcription termination by Pol II through interaction with CBC-ARS2 and NELF/DSIF, and promotes 3' end processing by enhancing recruitment of Integrator or Heat Labile Factor to snRNA or RDH genes, respectively.
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Benzonase® Nuclease, ultrapure, ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Triton X-100, laboratory grade