Merck
  • Evolutionary conserved NSL complex/BRD4 axis controls transcription activation via histone acetylation.
Evolutionary conserved NSL complex/BRD4 axis controls transcription activation via histone acetylation.
Nature communications (2020-05-10)
Aline Gaub, Bilal N Sheikh, M Felicia Basilicata, Marie Vincent, Mathilde Nizon, Cindy Colson, Matthew J Bird, James E Bradner, Julien Thevenon, Michael Boutros, Asifa Akhtar
ABSTRACT
Cells rely on a diverse repertoire of genes for maintaining homeostasis, but the transcriptional networks underlying their expression remain poorly understood. The MOF acetyltransferase-containing Non-Specific Lethal (NSL) complex is a broad transcription regulator. It is essential in Drosophila, and haploinsufficiency of the human KANSL1 subunit results in the Koolen-de Vries syndrome. Here, we perform a genome-wide RNAi screen and identify the BET protein BRD4 as an evolutionary conserved co-factor of the NSL complex. Using Drosophila and mouse embryonic stem cells, we characterise a recruitment hierarchy, where NSL-deposited histone acetylation enables BRD4 recruitment for transcription of constitutively active genes. Transcriptome analyses in Koolen-de Vries patient-derived fibroblasts reveals perturbations with a cellular homeostasis signature that are evoked by the NSL complex/BRD4 axis. We propose that BRD4 represents a conserved bridge between the NSL complex and transcription activation, and provide a new perspective in the understanding of their functions in healthy and diseased states.
Sigma-Aldrich
Anti-Mouse Serum antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-acetyl-Histone H4 (Lys16) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-KANSL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)