Merck
  • Allele-selective lowering of mutant HTT protein by HTT-LC3 linker compounds.
Allele-selective lowering of mutant HTT protein by HTT-LC3 linker compounds.
Nature (2019-11-02)
Zhaoyang Li, Cen Wang, Ziying Wang, Chenggang Zhu, Jie Li, Tian Sha, Lixiang Ma, Chao Gao, Yi Yang, Yimin Sun, Jian Wang, Xiaoli Sun, Chenqi Lu, Marian Difiglia, Yanai Mei, Chen Ding, Shouqing Luo, Yongjun Dang, Yu Ding, Yiyan Fei, Boxun Lu
ABSTRACT
Accumulation of mutant proteins is a major cause of many diseases (collectively called proteopathies), and lowering the level of these proteins can be useful for treatment of these diseases. We hypothesized that compounds that interact with both the autophagosome protein microtubule-associated protein 1A/1B light chain 3 (LC3)1 and the disease-causing protein may target the latter for autophagic clearance. Mutant huntingtin protein (mHTT) contains an expanded polyglutamine (polyQ) tract and causes Huntington's disease, an incurable neurodegenerative disorder2. Here, using small-molecule-microarray-based screening, we identified four compounds that interact with both LC3 and mHTT, but not with the wild-type HTT protein. Some of these compounds targeted mHTT to autophagosomes, reduced mHTT levels in an allele-selective manner, and rescued disease-relevant phenotypes in cells and in vivo in fly and mouse models of Huntington's disease. We further show that these compounds interact with the expanded polyQ stretch and could lower the level of mutant ataxin-3 (ATXN3), another disease-causing protein with an expanded polyQ tract3. This study presents candidate compounds for lowering mHTT and potentially other disease-causing proteins with polyQ expansions, demonstrating the concept of lowering levels of disease-causing proteins using autophagosome-tethering compounds.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
TEV Protease
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Polyglutamines antibody, Mouse monoclonal, ~2 mg/mL, clone 3B5H10, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8, ascites fluid, clone 1HU-4C8, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Spinocerebellar Ataxia Type 3 Antibody, clone 1H9, ascites fluid, clone 1H9, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Spectrin alpha chain (nonerythroid) Antibody, clone AA6, clone AA6, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
AN1, ≥98% (HPLC)
Pricing and availability is not currently available.