Merck
Integrative functional genomics decodes herpes simplex virus 1.
Nature communications (2020-04-29)
Adam W Whisnant, Christopher S Jürges, Thomas Hennig, Emanuel Wyler, Bhupesh Prusty, Andrzej J Rutkowski, Anne L'hernault, Lara Djakovic, Margarete Göbel, Kristina Döring, Jennifer Menegatti, Robin Antrobus, Nicholas J Matheson, Florian W H Künzig, Guido Mastrobuoni, Chris Bielow, Stefan Kempa, Chunguang Liang, Thomas Dandekar, Ralf Zimmer, Markus Landthaler, Friedrich Grässer, Paul J Lehner, Caroline C Friedel, Florian Erhard, Lars Dölken
ABSTRACT
The predicted 80 open reading frames (ORFs) of herpes simplex virus 1 (HSV-1) have been intensively studied for decades. Here, we unravel the complete viral transcriptome and translatome during lytic infection with base-pair resolution by computational integration of multi-omics data. We identify a total of 201 transcripts and 284 ORFs including all known and 46 novel large ORFs. This includes a so far unknown ORF in the locus deleted in the FDA-approved oncolytic virus Imlygic. Multiple transcript isoforms expressed from individual gene loci explain translation of the vast majority of ORFs as well as N-terminal extensions (NTEs) and truncations. We show that NTEs with non-canonical start codons govern the subcellular protein localization and packaging of key viral regulators and structural proteins. We extend the current nomenclature to include all viral gene products and provide a genome browser that visualizes all the obtained data from whole genome to single-nucleotide resolution.