Merck
  • Actin Assembly around the Shigella-Containing Vacuole Promotes Successful Infection.
Actin Assembly around the Shigella-Containing Vacuole Promotes Successful Infection.
Cell reports (2020-05-14)
Sonja Kühn, John Bergqvist, Magdalena Gil, Camila Valenzuela, Laura Barrio, Stéphanie Lebreton, Chiara Zurzolo, Jost Enninga
ABSTRACT
The enteroinvasive bacterium Shigella flexneri forces its uptake into non-phagocytic host cells through the translocation of T3SS effectors that subvert the actin cytoskeleton. Here, we report de novo actin polymerization after cellular entry around the bacterium-containing vacuole (BCV) leading to the formation of a dynamic actin cocoon. This cocoon is thicker than any described cellular actin structure and functions as a gatekeeper for the cytosolic access of the pathogen. Host CDC42, TOCA-1, N-WASP, WIP, the Arp2/3 complex, cortactin, coronin, and cofilin are recruited to the actin cocoon. They are subverted by T3SS effectors, such as IpgD, IpgB1, and IcsB. IcsB immobilizes components of the actin polymerization machinery at the BCV dependent on its fatty acyltransferase activity. This represents a unique microbial subversion strategy through localized entrapment of host actin regulators causing massive actin assembly. We propose that the cocoon promotes subsequent invasion steps for successful Shigella infection.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human SEPT7
Sigma-Aldrich
SMIFH2, ≥98% (HPLC)
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder
Sigma-Aldrich
Rac1 Inhibitor, Rac1 Inhibitor, CAS 1177865-17-6, is a cell-permeable, reversible inhibitor of Rac1 GDP/GTP exchange. Interferes with the interaction between Rac1 and Rac-specific GEFs Trio and Tiam1 (IC₅₀ ~50 µM).
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial