Merck
  • Generation of a conditional Flpo/FRT mouse model expressing constitutively active TGFβ in fibroblasts.
Generation of a conditional Flpo/FRT mouse model expressing constitutively active TGFβ in fibroblasts.
Scientific reports (2020-03-05)
Victoire Cardot-Ruffino, Véronique Chauvet, Cassandre Caligaris, Adrien Bertrand-Chapel, Nicolas Chuvin, Roxane M Pommier, Ulrich Valcourt, David F Vincent, Sylvie Martel, Sophie Aires, Bastien Kaniewski, Pierre Dubus, Philippe Cassier, Stéphanie Sentis, Laurent Bartholin
ABSTRACT
Transforming growth factor (TGFβ) is a secreted factor, which accumulates in tissues during many physio- and pathological processes such as embryonic development, wound healing, fibrosis and cancer. In order to analyze the effects of increased microenvironmental TGFβ concentration in vivo, we developed a conditional transgenic mouse model (Flpo/Frt system) expressing bioactive TGFβ in fibroblasts, a cell population present in the microenvironment of almost all tissues. To achieve this, we created the genetically-engineered [Fsp1-Flpo; FSFTGFβCA] mouse model. The Fsp1-Flpo allele consists in the Flpo recombinase under the control of the Fsp1 (fibroblast-specific promoter 1) promoter. The FSFTGFβCA allele consists in a transgene encoding a constitutively active mutant form of TGFβ (TGFβCA) under the control of a Frt-STOP-Frt (FSF) cassette. The FSFTGFβCA allele was created to generate this model, and functionally validated by in vitro, ex vivo and in vivo techniques. [Fsp1-Flpo; FSFTGFβCA] animals do not present any obvious phenotype despite the correct expression of TGFβCA transgene in fibroblasts. This [Fsp1-Flpo; FSFTGFβCA] model is highly pertinent for future studies on the effect of increased microenvironmental bioactive TGFβ concentrations in mice bearing Cre-dependent genetic alterations in other compartments (epithelial or immune compartments for instance). These dual recombinase system (DRS) approaches will enable scientists to study uncoupled spatiotemporal regulation of different genetic alterations within the same mouse, thus better replicating the complexity of human diseases.
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone 2-28-33, ascites fluid
Sigma-Aldrich
Guanidine thiocyanate, ≥97% (titration)
Roche
Collagenase D, from Clostridium histolyticum