Merck
  • Targeting Aquaporin-4 Subcellular Localization to Treat Central Nervous System Edema.
Targeting Aquaporin-4 Subcellular Localization to Treat Central Nervous System Edema.
Cell (2020-05-16)
Philip Kitchen, Mootaz M Salman, Andrea M Halsey, Charlotte Clarke-Bland, Justin A MacDonald, Hiroaki Ishida, Hans J Vogel, Sharif Almutiri, Ann Logan, Stefan Kreida, Tamim Al-Jubair, Julie Winkel Missel, Pontus Gourdon, Susanna Törnroth-Horsefield, Matthew T Conner, Zubair Ahmed, Alex C Conner, Roslyn M Bill
ABSTRACT
Swelling of the brain or spinal cord (CNS edema) affects millions of people every year. All potential pharmacological interventions have failed in clinical trials, meaning that symptom management is the only treatment option. The water channel protein aquaporin-4 (AQP4) is expressed in astrocytes and mediates water flux across the blood-brain and blood-spinal cord barriers. Here we show that AQP4 cell-surface abundance increases in response to hypoxia-induced cell swelling in a calmodulin-dependent manner. Calmodulin directly binds the AQP4 carboxyl terminus, causing a specific conformational change and driving AQP4 cell-surface localization. Inhibition of calmodulin in a rat spinal cord injury model with the licensed drug trifluoperazine inhibited AQP4 localization to the blood-spinal cord barrier, ablated CNS edema, and led to accelerated functional recovery compared with untreated animals. We propose that targeting the mechanism of calmodulin-mediated cell-surface localization of AQP4 is a viable strategy for development of CNS edema therapies.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
L-741,626, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥99.0% (TLC)
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
CelLytic M, Cell Lysis Reagent, Suitable for Mammalian cell lysis and protein solubilization.
Sigma-Aldrich
SIGMAFAST OPD, tablet
Sigma-Aldrich
Polyethylenimine, branched, average Mw ~25,000 by LS, average Mn ~10,000 by GPC, branched
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Trifluoperazine dihydrochloride, ≥99%, powder