Merck
  • Apatite germanates doped with tungsten: synthesis, structure, and conductivity.
Apatite germanates doped with tungsten: synthesis, structure, and conductivity.
Dalton transactions (Cambridge, England : 2003) (2010-10-05)
A Orera, T Baikie, E Kendrick, J F Shin, S Pramana, R Smith, T J White, M L Sanjuán, P R Slater
ABSTRACT
High oxygen content apatite germanates, La(10)Ge(6-x)W(x)O(27+x), have been prepared by doping on the Ge site with W. In addition to increasing the oxygen content, this doping strategy is shown to result in stabilisation of the hexagonal lattice, and yield high conductivities. Structural studies of La(10)Ge(5.5)W(0.5)O(27.5) show that the interstitial oxygen sites are associated to a different degree with the Ge/WO(4) tetrahedra, leading to five coordinate Ge/W and significant disorder for the oxygen sites associated with these units. Raman spectroscopy studies suggest that in the case of the WO(5) units, the interstitial oxygen is more tightly bonded and therefore not as mobile as in the case of the GeO(5) units, thus not contributing significantly to the conduction process.
Product Description
Supelco
Lanthanum(III) oxide, suitable for AAS, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Lanthanum(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Germanium(IV) oxide, (crystalline powder), 99.998% trace metals basis
Sigma-Aldrich
Germanium(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Germanium(IV) oxide, powder, 99.999% trace metals basis
Sigma-Aldrich
Lanthanum(III) oxide, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, nanopowder, <100 nm particle size (TEM), 99% trace metals basis