Merck
  • The cardenolides ouabain and reevesioside A promote FGF2 secretion and subsequent FGFR1 phosphorylation via converged ERK1/2 activation.
The cardenolides ouabain and reevesioside A promote FGF2 secretion and subsequent FGFR1 phosphorylation via converged ERK1/2 activation.
Biochemical pharmacology (2019-12-10)
Guan-Hao Zhao, Ya-Qi Qiu, Cheng-Wei Yang, Ih-Sheng Chen, Chin-Yu Chen, Shiow-Ju Lee
ABSTRACT
Na+/K+-ATPase α1 was reported to directly interact with and recruit FGF2 (fibroblast growth factor 2), a vital cell signaling protein implicated in angiogenesis, to the inner plasma membrane for subsequent secretion. Cardenolides, a class of cardiac glycosides, were reported to downregulate FGF2 secretion upon binding to Na+/K+-ATPase α1 in a cell system with ectopically expressed FGF2 and Na+/K+-ATPase α1. Herein, we disclose that the cardenolides ouabain and reevesioside A significantly enhance the secretion/release of FGF2 and the phosphorylation of FGFR1 (fibroblast growth factor receptor 1) in a time- and dose-dependent manner, in A549 carcinoma cells. A pharmacological approach was used to elucidate the pertinent upstream effectors. Only the ERK1/2 inhibitor U0126 but not the other inhibitors examined (including those inhibiting the unconventional secretion of FGF2) was able to reduce ouabain-induced FGF2 secretion and FGFR1 activation. ERK1/2 phosphorylation was increased upon ouabain treatment, a process found to be mediated through upstream effectors including ouabain-induced phosphorylated EGFR and a reduced MKP1 protein level. Therefore, at least two independent lines of upstream effectors are able to mediate ouabain-induced ERK1/2 phosphorylation and the subsequent FGF2 secretion and FGFR1 activation. These finding constitute unprecedent insights into the regulation of FGF2 secretion by cardenolides.
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
LFM-A13, ≥98% (HPLC), powder
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Ouabain, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Supelco
Digoxin, analytical standard
Sigma-Aldrich
NSC 95397, ≥97% (HPLC)
Sigma-Aldrich
Ouabain octahydrate, ≥95% (HPLC), powder