Merck
A 6-nm ultra-photostable DNA FluoroCube for fluorescence imaging.
Nature methods (2020-03-24)
Stefan Niekamp, Nico Stuurman, Ronald D Vale
ABSTRACT
Photobleaching limits extended imaging of fluorescent biological samples. We developed DNA-based 'FluoroCubes' that are similar in size to the green fluorescent protein, have single-point attachment to proteins, have a ~54-fold higher photobleaching lifetime and emit ~43-fold more photons than single organic dyes. We demonstrate that DNA FluoroCubes provide outstanding tools for single-molecule imaging, allowing the tracking of single motor proteins for >800 steps with nanometer precision.
Sigma-Aldrich
Pyruvate Kinase/Lactic Dehydrogenase enzymes from rabbit muscle, For the Determination of ADP, buffered aqueous glycerol solution
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Phospho(enol)pyruvic acid monopotassium salt, 99%
Sigma-Aldrich
β-Casein from bovine milk, BioUltra, ≥98% (PAGE)
Sigma-Aldrich
κ-Casein from bovine milk, ≥70% (PAGE), lyophilized powder