Merck
  • A Pan-plant Protein Complex Map Reveals Deep Conservation and Novel Assemblies.
A Pan-plant Protein Complex Map Reveals Deep Conservation and Novel Assemblies.
Cell (2020-03-20)
Claire D McWhite, Ophelia Papoulas, Kevin Drew, Rachael M Cox, Viviana June, Oliver Xiaoou Dong, Taejoon Kwon, Cuihong Wan, Mari L Salmi, Stanley J Roux, Karen S Browning, Z Jeffrey Chen, Pamela C Ronald, Edward M Marcotte
ABSTRACT
Plants are foundational for global ecological and economic systems, but most plant proteins remain uncharacterized. Protein interaction networks often suggest protein functions and open new avenues to characterize genes and proteins. We therefore systematically determined protein complexes from 13 plant species of scientific and agricultural importance, greatly expanding the known repertoire of stable protein complexes in plants. By using co-fractionation mass spectrometry, we recovered known complexes, confirmed complexes predicted to occur in plants, and identified previously unknown interactions conserved over 1.1 billion years of green plant evolution. Several novel complexes are involved in vernalization and pathogen defense, traits critical for agriculture. We also observed plant analogs of animal complexes with distinct molecular assemblies, including a megadalton-scale tRNA multi-synthetase complex. The resulting map offers a cross-species view of conserved, stable protein assemblies shared across plant cells and provides a mechanistic, biochemical framework for interpreting plant genetics and mutant phenotypes.
Sigma-Aldrich
Gel Filtration Markers Kit for Protein Molecular Weights 29,000-700,000 Da
Sigma-Aldrich
Bovine Serum Albumin, pH <5.0, powder
Sigma-Aldrich
β-Amylase from sweet potato
Sigma-Aldrich
Thyroglobulin from bovine thyroid, (For Gel Filtration Chromatography)