Merck
  • Preclinical Activity of Ribociclib in Squamous Cell Carcinoma of the Head and Neck.
Preclinical Activity of Ribociclib in Squamous Cell Carcinoma of the Head and Neck.
Molecular cancer therapeutics (2020-01-12)
Gabrielle van Caloen, Sandra Schmitz, Mariama El Baroudi, Xavier Caignet, Sébastien Pyr Dit Ruys, Pierre P Roger, Didier Vertommen, Jean-Pascal Machiels
ABSTRACT
Cell-cycle pathway impairments resulting in CDK4 and 6 activation are frequently observed in human papillomavirus (HPV)-negative squamous cell carcinoma of the head and neck (SCCHN). We investigated the activity of ribociclib, a CDK4/6 inhibitor, in SCCHN models with the aim of identifying predictive biomarkers of response. HPV-negative or HPV-positive SCCHN cell lines (n = 8) and patient-derived tumor xenograft (PDTX) models (n = 6) were used. The models were classified according to their sensitivity to ribociclib to investigate potential predictive biomarkers. Ribociclib had a cytostatic effect in some HPV-negative SCCHN models but had no effect in HPV-positive models. In SCCHN cell lines and PDTXs, the retinoblastoma (Rb) protein expression level correlated with ribociclib activity. Rb knockdown was, however, not sufficient to block G0-G1 arrest induced by ribociclib in Detroit-562 where p107, p130, and Forkhead BOX M1 (FOXM1) were also implicated in ribociclib activity. Cell lines harboring epithelial-to-mesenchymal transition (EMT) features were less sensitive to ribociclib than those with an epithelial phenotype. Rb downregulation induced EMT in our Rb-expressing SCCHN cell lines. However, ribociclib still had significant activity in one PDTX model with high Rb and vimentin expression, suggesting that the presence of vimentin alone is not enough to induce ribociclib resistance. These findings suggest that CDK4/6 inhibitors should be investigated in patients with HPV-negative SCCHN with high Rb expression and an epithelial phenotype. Although these biomarkers are not predictive in all cases, they may enrich the population that could benefit from CDK4/6 inhibitors.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® esiRNA, targeting human RB1
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution