Merck
PBX1 expression in uterine natural killer cells drives fetal growth.
Science translational medicine (2020-04-03)
Yonggang Zhou, Binqing Fu, Xiuxiu Xu, Jinghe Zhang, Xianhong Tong, Yanshi Wang, Zhongjun Dong, Xiaoren Zhang, Nan Shen, Yiwen Zhai, Xiangdong Kong, Rui Sun, Zhigang Tian, Haiming Wei
ABSTRACT
Abundant decidual natural killer (dNK) cells at the maternal-fetal interface are important during early pregnancy. However, functional subsets of dNK cells remain poorly understood. We describe a CD49a+PBX homeobox 1 (PBX1)+ dNK cell subset that promotes fetal development in humans and mice. The expression of PBX1 in dNK cells is up-regulated via the activated AKT1 pathway through the interaction of major histocompatibility complex G with the immunoglobulin-like transcript 2 receptor. PBX1 drives pleiotrophin and osteoglycin transcription in dNK cells, further promoting fetal development. Decreased PBX1 expression or the PBX1G21S mutant correlated with fetal growth restriction and pregnancy failure in patients with unexplained recurrent spontaneous abortion (URSA). Inactivation of Pbx1 in mouse dNK cells impairs fetal development by decreasing growth-promoting factors from CD49a+PBX1+ dNK cells. Impairment of PBX1 in dNK cells has positive correlation with URSA pathogenesis and may provide a potential marker for this condition.
Sigma-Aldrich
MISSION® esiRNA, targeting human PDK2
Sigma-Aldrich
MISSION® esiRNA, targeting human AKT1, RP11-982M15.2
Sigma-Aldrich
MISSION® esiRNA, targeting human PBX1
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1