Merck
  • The exosomal integrin α5β1/AEP complex derived from epithelial ovarian cancer cells promotes peritoneal metastasis through regulating mesothelial cell proliferation and migration.
The exosomal integrin α5β1/AEP complex derived from epithelial ovarian cancer cells promotes peritoneal metastasis through regulating mesothelial cell proliferation and migration.
Cellular oncology (Dordrecht) (2020-02-23)
Xiaoduan Li, Meiling Tang, Qinyi Zhu, Xinjing Wang, Yingying Lin, Xipeng Wang
ABSTRACT
Epithelial ovarian cancer (EOC) is one of the most malignant cancers in the gynecologic system. Many patients are diagnosed at an advanced stage with disseminated intra-peritoneal metastases. EOC spreads via both direct extension and trans-coelomic spread. However, the interplay between human peritoneal mesothelial cells (HPMCs) and EOC cells is still ambiguous. We hypothesize that integrins (ITG) in HPMCs may play important roles in EOC metastasis. The expression of different integrin subtypes from HPMCs was assessed using Western blotting. The expression of integrin α5β1 (ITGA5B1) and its co-localization with asparaginyl endopeptidase (AEP) in HPMCs derived from EOC patients (EOC-HPMCs) were assessed using immunofluorescence. The role and mechanism of the exosomal ITGA5B1/AEP complex in HPMCs was assessed using both in vitro and in vivo assays. A retrospective study involving 234 cases was carried out to assess ITGA5B1 and AEP levels in circulating sera and ascites of EOC patients, as well as associations between ITGA5B1/AEP expression and overall survival. We found that ITGA5B1was highly expressed and co-localized with AEP in EOC cells, and that the exosomal ITGA5B1/AEP complex secreted by EOC cells played an important role in the proliferation and migration of HPMCs. High levels of exosomal ITGA5B1/AEP were also found in circulating sera and ascites of EOC patients, and the expression of ITGA5B1/AEP in EOC tissues was found to be negatively associated with overall survival. Our data indicate that EOCs may regulate the function of HPMCs through exosomal ITGA5B1/AEP, which may be crucial for peritoneal metastasis.
Product Description
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
Human LGMN / Legumain ELISA Kit