Merck
  • Rabies virus glycoprotein-amplified hierarchical targeted hybrids capable of magneto-electric penetration delivery to orthotopic brain tumor.
Rabies virus glycoprotein-amplified hierarchical targeted hybrids capable of magneto-electric penetration delivery to orthotopic brain tumor.
Journal of controlled release : official journal of the Controlled Release Society (2020-02-12)
Yu-Lin Su, Li-Wen Kuo, Chia-Hsien Hsu, Chi-Shiun Chiang, Yu-Jen Lu, Shing-Jyh Chang, Shang-Hsiu Hu
ABSTRACT
Compact nanohybrids can potentially unite various therapeutic features and reduce side effects for precise cancer therapy. However, the poor accumulation and limited tumor penetration of drugs at the tumor impede the manifestation of nanomedicine. We developed a rabies virus glycoprotein (RVG)-amplified hierarchical targeted hybrid that acts as a stealthy and magnetolytic carrier that transports dual tumor-penetrating agents incorporating two drugs (boron-doped graphene quantum dots (B-GQDs)/doxorubicin and pH-responsive dendrimers (pH-Den)/palbociclib). The developed RVG-decorated hybrids (RVG-hybrids) enhance the accumulation of drugs at tumor by partially bypassing the BBB via spinal cord transportation and pH-induced aggregation of hierarchical targeting. The penetrated delivery of dual pH-Den and B-GQD drugs to deep tumors is actuated by magnetoelectric effect, which are able to generate electrons to achieve electrostatic repulsion and disassemble the hybrids into components of a few nanometers in size. The synergy of magnetoelectric drug penetration and chemotherapy was achieved by delivery of the B-GQDs and pH-Den to orthotopic tumors, which prolonged the host survival time. This RVG-amplified dual hierarchical delivery integrated with controlled and penetrated release from this hybrid improve the distribution of the therapeutic agents at the brain tumor for synergistic therapy, exhibiting potential for clinic use.
Sigma-Aldrich
4-(Bromomethyl)phenylboronic acid
Pricing and availability is not currently available.