Merck
  • Metalloproteinase PAPP-A regulation of IGF-1 contributes to polycystic kidney disease pathogenesis.
Metalloproteinase PAPP-A regulation of IGF-1 contributes to polycystic kidney disease pathogenesis.
JCI insight (2020-01-29)
Sonu Kashyap, Kyaw Zaw Hein, Claudia Cs Chini, Jorgo Lika, Gina M Warner, Laurie K Bale, Vicente E Torres, Peter C Harris, Claus Oxvig, Cheryl A Conover, Eduardo N Chini
ABSTRACT
Autosomal dominant polycystic kidney disease (ADPKD) is the most common genetic cause of end-stage renal disease (ESRD). The treatment options for ADPKD are limited. We observed an upregulation in several IGF-1 pathway genes in the kidney of Pkd1RC/RC mice, a model of ADPKD. Pregnancy-associated plasma protein A (PAPP-A), a metalloproteinase that cleaves inhibitory IGF binding proteins (IGFBPs), increasing the local bioactivity of IGF-1, was highly induced in the kidney of ADPKD mice. PAPP-A levels were high in cystic fluid and kidneys of humans with ADPKD. Our studies further showed that PAPP-A transcription in ADPKD was mainly regulated through the cAMP/CREB/CBP/p300 pathway. Pappa deficiency effectively inhibited the development of cysts in the Pkd1RC/RC mice. The role of PAPP-A in cystic disease appears to be regulation of the IGF-1 pathway and cellular proliferation in the kidney. Finally, preclinical studies demonstrated that treatment with a monoclonal antibody that blocks the proteolytic activity of PAPP-A against IGFBP4 ameliorated ADPKD cystic disease in vivo in Pkd1RC/RC mice and ex vivo in embryonic kidneys. These data indicated that the PAPP-A/IGF-1 pathway plays an important role in the growth and expansion of cysts in ADPKD. Our findings introduce a therapeutic strategy for ADPKD that involves the inhibition of PAPP-A.
Sigma-Aldrich
Human PAPPA / Pappalysin-1 ELISA Kit