Merck
  • Versican, a major hyaluronan-binding component in the dermis, loses its hyaluronan-binding ability in solar elastosis.
Versican, a major hyaluronan-binding component in the dermis, loses its hyaluronan-binding ability in solar elastosis.
The Journal of investigative dermatology (2007-03-17)
Keiko Hasegawa, Masahiko Yoneda, Hiroko Kuwabara, Osamu Miyaishi, Naoki Itano, Akiko Ohno, Masahiro Zako, Zenzo Isogai
ABSTRACT
Versican interacts with hyaluronan (HA) at its N-terminus and with fibrillin-1 at its C terminus. As versican in the dermis connects microfibrils to the HA-rich matrix for viscoelasticity, dermal diseases may involve destruction of these complexes. A recombinant versican protein, rVN, covering the HA binding region (HABR) of human versican and a polyclonal antibody, 6084, against rVN were prepared and characterized. Blotting analyses of skin extracts with 6084 and biotin-conjugated HA revealed that versican was a major HA-binding component in the dermis. Matrix metalloprotease-12, which is expressed in areas of solar elastosis, degraded versican and abrogated its HA-binding ability. Immunohistochemical analyses revealed that the elastic materials in solar elastosis lesions were negative for 6084, but positive for 2B1, an antibody recognizing the C-terminus of versican, indicating loss of the HABR in the aggregated elastic fibers. This loss of the HA-binding ability of versican followed by HA exclusion may be responsible for the pathological and phenotypical changes observed in solar elastosis.
Sigma-Aldrich
Hyaluronan biotin sodium salt, ≥97%, soluble powder