Merck
Parallel lipoplex folding pathways revealed using magnetic tweezers.
Biomacromolecules (2012-09-20)
Zhiqiang Sun, Elena B Tikhonova, Helen I Zgurskaya, Valentin V Rybenkov
ABSTRACT
Lipid-coated DNA nanoparticles (lipoplexes) are a powerful gene delivery tool with promising therapeutic applications. The mechanism of lipoplex assembly remains poorly understood. We explored DNA packing by a cationic lipid DSTAP (distearoyl trimethylammonium-propane) using magnetic tweezers. DSTAP-induced DNA condensation occurred as a series of bursts with the mean step size of 60-80 nm. The pause time preceding the steps could be approximated as a bimodal distribution, which reveals at least two distinct condensation pathways. The rapidly condensed DNA was more resilient to force-induced decondensation. The proportion of the stable, fast-formed complexes decreased at high salt concentrations. A similar trend was observed in bulk experiments. Lipoplexes assembled at low salt concentration more efficiently shielded DNA from fluorescent dyes and DNase even after transfer to the high salt conditions. These data reveal that lipoplex folding occurs via two parallel pathways even at the single molecule level. The progress through the two pathways can be monitored in real time using single DNA manipulations. The relative efficiency of the two pathways can be varied by external conditions.
Product Description
Avanti
18:0 TAP, Avanti Research - A Croda Brand