Merck
  • 7beta-hydroxycholesterol induces Ca(2+) oscillations, MAP kinase activation and apoptosis in human aortic smooth muscle cells.
7beta-hydroxycholesterol induces Ca(2+) oscillations, MAP kinase activation and apoptosis in human aortic smooth muscle cells.
Atherosclerosis (2000-11-04)
M P Ares, M I Pörn-Ares, S Moses, J Thyberg, L Juntti-Berggren, P Berggren, A Hultgårdh-Nilsson, B Kallin, J Nilsson
ABSTRACT
In the present study, we characterize the early cytotoxic effects of 7beta-hydroxycholesterol, a major cytotoxin in oxidized LDL, in human aortic smooth muscle cells. Within a few minutes after addition, 7beta-hydroxycholesterol induced Ca(2+) oscillations with a frequency of approximately 0.3-0.4 min(-1). A few hours later, thapsigargin-sensitive Ca(2+) pools were depleted, indicating that 7beta-hydroxycholesterol perturbs intracellular Ca(2+) homeostasis. The mitogen-activated protein kinases (MAPKs) ERK1 and ERK2 (but not JNK) were activated within 5 min after addition of 7beta-hydroxycholesterol. The side-chain hydroxylated oxysterols 25-hydroxycholesterol and 27-hydroxycholesterol were more potent in inducing apoptosis than 7beta-hydroxycholesterol and cholesterol-5alpha,6alpha-epoxide, as determined by TUNEL staining. Addition of TNFalpha (10 ng/ml) and IFNgamma (20 ng/ml) enhanced the cytotoxicity of oxysterols and potentiated apoptosis. The cytokines alone were not toxic to smooth muscle cells at these concentrations. 25-Hydroxycholesterol and 7beta-hydroxycholesterol but not cholesterol inhibited protein synthesis at 4-8 h as determined by [35S]methionine incorporation assay. Morphologically, oxysterol-induced cell death was characterized by disorganization of the ER and Golgi membranes. The Ca(2+) and ERK signals preceded the ultrastructural changes induced by 7beta-hydroxycholesterol.
Product Description
Avanti
7β-hydroxycholesterol, Avanti Research - A Croda Brand