Merck
  • Cytochrome P450 oxidoreductase contributes to phospholipid peroxidation in ferroptosis.
Cytochrome P450 oxidoreductase contributes to phospholipid peroxidation in ferroptosis.
Nature chemical biology (2020-02-23)
Yilong Zou, Haoxin Li, Emily T Graham, Amy A Deik, John K Eaton, Wenyu Wang, Gerardo Sandoval-Gomez, Clary B Clish, John G Doench, Stuart L Schreiber
ABSTRACT
Ferroptosis is widely involved in degenerative diseases in various tissues including kidney, liver and brain, and is a targetable vulnerability in multiple primary and therapy-resistant cancers. Accumulation of phospholipid hydroperoxides in cellular membranes is the hallmark and rate-limiting step of ferroptosis; however, the enzymes contributing to lipid peroxidation remain poorly characterized. Using genome-wide, CRISPR-Cas9-mediated suppressor screens, we identify cytochrome P450 oxidoreductase (POR) as necessary for ferroptotic cell death in cancer cells exhibiting inherent and induced susceptibility to ferroptosis. By genetic depletion of POR in cancer cells, we reveal that POR contributes to ferroptosis across a wide range of lineages and cell states, and in response to distinct mechanisms of ferroptosis induction. Using systematic lipidomic profiling, we further map POR's activity to the lipid peroxidation step in ferroptosis. Hence, our work suggests that POR is a key mediator of ferroptosis and potential druggable target for developing antiferroptosis therapeutics.
Sigma-Aldrich
1S,3R-RSL 3, ≥98% (HPLC)
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)
Sigma-Aldrich
Erastin, ≥98% (HPLC)
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
2-(Diisopropylamino)ethyl methacrylate, 97%, contains ~100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
Sigma-Aldrich
ML 210, ≥98% (HPLC)
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%