Merck
  • A truncating mutation in the autophagy gene UVRAG drives inflammation and tumorigenesis in mice.
A truncating mutation in the autophagy gene UVRAG drives inflammation and tumorigenesis in mice.
Nature communications (2019-12-14)
Christine Quach, Ying Song, Hongrui Guo, Shun Li, Hadi Maazi, Marshall Fung, Nathaniel Sands, Douglas O'Connell, Sara Restrepo-Vassalli, Billy Chai, Dali Nemecio, Vasu Punj, Omid Akbari, Gregory E Idos, Shannon M Mumenthaler, Nancy Wu, Sue Ellen Martin, Ashley Hagiya, James Hicks, Hengmin Cui, Chengyu Liang
ABSTRACT
Aberrant autophagy is a major risk factor for inflammatory diseases and cancer. However, the genetic basis and underlying mechanisms are less established. UVRAG is a tumor suppressor candidate involved in autophagy, which is truncated in cancers by a frameshift (FS) mutation and expressed as a shortened UVRAGFS. To investigate the role of UVRAGFS in vivo, we generated mutant mice that inducibly express UVRAGFS (iUVRAGFS). These mice are normal in basal autophagy but deficient in starvation- and LPS-induced autophagy by disruption of the UVRAG-autophagy complex. iUVRAGFS mice display increased inflammatory response in sepsis, intestinal colitis, and colitis-associated cancer development through NLRP3-inflammasome hyperactivation. Moreover, iUVRAGFS mice show enhanced spontaneous tumorigenesis related to age-related autophagy suppression, resultant β-catenin stabilization, and centrosome amplification. Thus, UVRAG is a crucial autophagy regulator in vivo, and autophagy promotion may help prevent/treat inflammatory disease and cancer in susceptible individuals.
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
TRI Reagent®, BD, For processing whole blood, plasma, or serum.
Sigma-Aldrich
Azoxymethane, 13.4 M, ≥98%
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
iCRT14, ≥98% (HPLC)
Sigma-Aldrich
Anti-phospho-UVRAG (Ser498), from rabbit