Merck
  • Influence of Forest Disturbance on La Crosse Virus Risk in Southwestern Virginia.
Influence of Forest Disturbance on La Crosse Virus Risk in Southwestern Virginia.
Insects (2020-01-08)
M Camille Hopkins, Steven D Zink, Sally L Paulson, Dana M Hawley
ABSTRACT
Forest disturbance effects on La Crosse virus (LACV) are currently unknown. We determined the abundance of three LACV accessory vectors (Aedes albopictus, Ae. canadensis, and Ae. vexans) and the primary amplifying host (Eastern chipmunk; Tamias striatus), and tested for LACV prevalence in both vectors and chipmunks, across a gradient of experimental forest disturbance treatments in southwest Virginia. Forest disturbance significantly affected the abundance of LACV accessory vectors, with a higher abundance on disturbed sites for Ae. canadensis and Ae.vexans. However, there was no significant disturbance effect on chipmunk abundance. Forest disturbance significantly affected LACV prevalence in mosquito vectors, with most (80%) detections on unlogged control sites, which past work showed harbor the highest abundance of the two most common LACV vectors (the primary vector Aedes triseriatus, and Ae. japonicus). Interestingly, LACV nucleic acid was only detected in Ae. japonicus and Culex pipiens/restuans, with no detections in the primary vector, Ae. triseriatus. In contrast to the vector results, antibodies were only found in chipmunks on logged sites, but this result was not statistically significant. Overall, our results suggest that human LACV risk should generally decline with logging, and reveal the potential importance of accessory vectors in LACV maintenance in Appalachian forests.
Sigma-Aldrich
Mouse serum
Sigma-Aldrich
Complement sera from guinea pig, lyophilized powder