Merck
  • Deregulated MTOR (mechanistic target of rapamycin kinase) is responsible for autophagy defects exacerbating kidney stone development.
Deregulated MTOR (mechanistic target of rapamycin kinase) is responsible for autophagy defects exacerbating kidney stone development.
Autophagy (2019-07-02)
Rei Unno, Tsuyoshi Kawabata, Kazumi Taguchi, Teruaki Sugino, Shuzo Hamamoto, Ryosuke Ando, Atsushi Okada, Kenjiro Kohri, Tamotsu Yoshimori, Takahiro Yasui
ABSTRACT
Kidney stone disease is a lifestyle-related disease prevalent in developed countries; however, effective medical treatment for the disease is not yet well established. As cellular damage in renal tubular cells (RTCs) is responsible for the disease, here, we focused on the role of macroautophagy/autophagy in RTCs. We found that autophagic activity was significantly decreased in mouse RTCs exposed to calcium oxalate (CaOx) monohydrate crystals and in the kidneys of GFP-conjugated MAP1LC3B (microtubule- associated protein 1 light chain 3 beta) transgenic mice with CaOx nephrocalcinosis induced by glyoxylate. This caused accumulation of damaged intracellular organelles, such as mitochondria and lysosomes, the normal functioning of which is mediated by functional autophagy. An impairment of autophagy was also observed in the mucosa with plaques of CaOx kidney stone formers. We determined that the decrease in autophagy was caused by an upregulation of MTOR (mechanistic target of rapamycin kinase), which consequently resulted in the suppression of the upstream autophagy regulator TFEB (transcription factor EB). Furthermore, we showed that an MTOR inhibitor could recover a decrease in autophagy and alleviate crystal-cell interactions and the formation of crystals associated with increased inflammatory responses. Taken together, we conclude that autophagy compromised by MTOR deregulation is a fundamental feature in the pathology of kidney stone formation, and propose that chemical inhibition of MTOR could be a prospective strategy for disease suppression.Abbreviations: ACTB: actin, beta; CaOx: calcium oxalate; CKD: chronic kidney disease; COM: calcium oxalate monohydrate; LGALS3/galectin-3: lectin, galactose binding, soluble 3; GFP: green fluorescent protein; GOX: glyoxylate; HE: hematoxylin and eosin; MAPLC3B: microtubule- associated protein 1 light chain 3 beta; MTOR: mechanistic target of rapamycin kinase; NAC: N-acetyl-L-cysteine; ROS: reactive oxygen species; RTC: renal tubular cell; SQSTM1/p62: sequestosome 1; TFEB: transcription factor EB; TEM: transmission electron microscopy; tfLC3: tandem fluorescent-tagged LC3; 3-MA: 3-methyladenine.
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Earle′s Balanced Salt Solution 10x, Without sodium bicarbonate, 10 ×, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, suitable for cell culture, BioReagent
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-Thiazide-Sensitive NaCl Cotransporter Antibody, Chemicon®, from rabbit
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Leu-Leu methyl ester hydrobromide, ≥97% (TLC)
Sigma-Aldrich
L-Ascorbic acid, 99%