Merck
  • Dissociated Hippocampal Neurons Exhibit Distinct Zn2+ Dynamics in a Stimulation-Method-Dependent Manner.
Dissociated Hippocampal Neurons Exhibit Distinct Zn2+ Dynamics in a Stimulation-Method-Dependent Manner.
ACS chemical neuroscience (2020-02-06)
Lynn Sanford, Amy E Palmer
ABSTRACT
Ionic Zn2+ has increasingly been recognized as an important neurotransmitter and signaling ion in glutamatergic neuron pathways. Intracellular Zn2+ transiently increases as a result of neuronal excitation, and this Zn2+ signal is essential for neuron plasticity, but the source and regulation of the signal is still unclear. In this study, we rigorously quantified Zn2+, Ca2+, and pH dynamics in dissociated mouse hippocampal neurons stimulated with bath application of high KCl or glutamate. While both stimulation methods yielded Zn2+ signals, Ca2+ influx, and acidification, glutamate stimulation induced more sustained high intracellular Ca2+ and a larger increase in intracellular Zn2+. However, the stimulation-induced pH change was similar between conditions, indicating that a different cellular change is responsible for the stimulation-dependent difference in Zn2+ signal. This work provides the first robust quantification of Zn2+ dynamics in neurons using different methods of stimulation.
Sigma-Aldrich
Chelex® 100 sodium form, 50-100 mesh (dry)
Sigma-Aldrich
1-Hydroxy-2(1H)-pyridinethione, ≥95%
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
2-Mercaptopyridine N-oxide, 99%
Sigma-Aldrich
2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein, ≥90% (HPLC)
Sigma-Aldrich
Nigericin sodium salt, ≥98% (TLC)
Sigma-Aldrich
Ionomycin, Free Acid, Streptomyces conglobatus, Ionomycin, Free Acid, Streptomyces conglobatus, CAS 56092-81-0, is a highly specific for divalent cations and more effective than A23187 (Cat. No. 100105) as a mobile ion carrier for Ca2+.
Sigma-Aldrich
Calcium chloride, desiccant, ACS reagent, ≥96.0%