Merck
Snake Venom Gland Organoids.
Cell (2020-01-25)
Yorick Post, Jens Puschhof, Joep Beumer, Harald M Kerkkamp, Merijn A G de Bakker, Julien Slagboom, Buys de Barbanson, Nienke R Wevers, Xandor M Spijkers, Thomas Olivier, Taline D Kazandjian, Stuart Ainsworth, Carmen Lopez Iglesias, Willine J van de Wetering, Maria C Heinz, Ravian L van Ineveld, Regina G D M van Kleef, Harry Begthel, Jeroen Korving, Yotam E Bar-Ephraim, Walter Getreuer, Anne C Rios, Remco H S Westerink, Hugo J G Snippert, Alexander van Oudenaarden, Peter J Peters, Freek J Vonk, Jeroen Kool, Michael K Richardson, Nicholas R Casewell, Hans Clevers
ABSTRACT
Wnt dependency and Lgr5 expression define multiple mammalian epithelial stem cell types. Under defined growth factor conditions, such adult stem cells (ASCs) grow as 3D organoids that recapitulate essential features of the pertinent epithelium. Here, we establish long-term expanding venom gland organoids from several snake species. The newly assembled transcriptome of the Cape coral snake reveals that organoids express high levels of toxin transcripts. Single-cell RNA sequencing of both organoids and primary tissue identifies distinct venom-expressing cell types as well as proliferative cells expressing homologs of known mammalian stem cell markers. A hard-wired regional heterogeneity in the expression of individual venom components is maintained in organoid cultures. Harvested venom peptides reflect crude venom composition and display biological activity. This study extends organoid technology to reptilian tissues and describes an experimentally tractable model system representing the snake venom gland.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Neutral Red Solution (0.33%), 3.3 g/L in DPBS, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 647 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
Sigma-Aldrich
Gentamicin solution, 50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade