Merck
  • Synthesis of an Alkynyl Methylglyoxal Probe to Investigate Nonenzymatic Histone Glycation.
Synthesis of an Alkynyl Methylglyoxal Probe to Investigate Nonenzymatic Histone Glycation.
The Journal of organic chemistry (2019-12-26)
Qingfei Zheng, Igor Maksimovic, Akhil Upad, David Guber, Yael David
ABSTRACT
Methylglyoxal (MGO) is a reactive dicarbonyl metabolite that modifies histones in vivo and induces changes in chromatin structure and function. Here we report the synthesis and application of a chemical probe for investigating MGO-glycation. A two-step synthesis of a Cu-click compatible alkynyl oxoaldehyde probe (AlkMGO) via sequential Dess-Martin and Riley oxidations is presented. This synthesis elevates the accessibility and utility of an important tool for tracking, enriching, and studying MGO-glycation to aid in understanding its underlying biochemical functions.
Sigma-Aldrich
Cy5-azide, 95% (HPLC)
Sigma-Aldrich
Azide-PEG3-biotin conjugate
Sigma-Aldrich
N-(3-Azidopropyl)biotinamide, ≥95%