Merck
  • Characterization of bradykinin B2 receptors on human IMR-90 lung fibroblasts: stimulation of 45Ca2+ efflux by D-Phe substituted bradykinin analogues.
Characterization of bradykinin B2 receptors on human IMR-90 lung fibroblasts: stimulation of 45Ca2+ efflux by D-Phe substituted bradykinin analogues.
European journal of pharmacology (1992-11-02)
D G Sawutz, D M Faunce, W T Houck, D Haycock
ABSTRACT
[3H]Bradykinin binds to intact human IMR-90 fetal lung fibroblasts in a time and dose-dependent manner. Binding equilibrium was attained by 120 minutes at 4 degrees C. [3H]Bradykinin binding was saturable; Scatchard analysis of saturation binding data demonstrated a single binding site having a KD = 1.8 +/- 0.2 nM and a receptor concentration of 17.4 +/- 4.0 fmol/10(5) cells. The calculated value for KD(k-1/k1) from the association (k1 = 4.71 x 10(6) mol-1 min-1) and dissociation (k-1 = 1.13 x 10(-2) min-1) rate constants was 2.4 nM. The rank order of potency observed for bradykinin peptide agonists, bradykinin > Lys-bradykinin > Met,Lys-bradykinin > Ile,Ser-bradykinin > des-Arg9-bradykinin, is consistent with that of a bradykinin B2 receptor. Bradykinin stimulated efflux of 45Ca2+ from IMR-90 cells dose dependently with an EC50 = 331 +/- 50 pM. 45Ca2+ efflux was also demonstrated with Lys-bradykinin and Met-Lys-bradykinin but not by des-Arg10-kallidin (100 nM) or NKA (1 microM). Hoe-140 inhibited bradykinin-induced 45Ca2+ efflux (IC50 = 3 +/- 2 nM). D-Phe7-substituted bradykinin analogues stimulated 45Ca2+ efflux dose dependently and this stimulation of 45Ca2+ efflux was inhibited by Hoe-140. These results suggest that D-Phe7 substituted bradykinin analogues are agonists at the bradykinin B2 receptor in IMR-90 cells.
Sigma-Aldrich
Bradykinin Ile-Ser, T-Kinin