Merck
  • Label-Free Interactome Analysis Revealed an Essential Role of CUL3-KEAP1 Complex in Mediating the Ubiquitination and Degradation of PHD2.
Label-Free Interactome Analysis Revealed an Essential Role of CUL3-KEAP1 Complex in Mediating the Ubiquitination and Degradation of PHD2.
Journal of proteome research (2019-11-26)
Ang Luo, Yue Chen
ABSTRACT
Prolyl hydroxylase domain-containing protein 2 (PHD2/EGLN1) is a key regulatory enzyme that plays a fundamental role in the cellular hypoxic response pathway, mediating proline hydroxylation-dependent protein degradation of selected target proteins. However, the regulation of PHD2 homeostasis at the protein level is not well understood. Here, we perform label-free quantitative interactome analysis through immunoprecipitation coupled with mass spectrometry analysis. To minimize the side effects caused by ectopic overexpression, in HeLa cells, we stably overexpressed Flag-tagged PHD2 while suppressing the endogenous PHD2 by using an shRNA targeting its 3' UTR region. We identified and validated Cullin 3 as a novel PHD2 interactor in vivo. Through candidate screening, we further identified CUL3-KEAP1 E3 ubiquitin ligase complex as the major enzyme that regulates PHD2 degradation. Overexpression of either CUL3, KEAP1, or both significantly increases PHD2 ubiquitination and reduces PHD2 protein abundance. The knockdown of CUL3 or KEAP1 decreased PHD2 ubiquitination and inhibited PHD2 degradation. Accordingly, loss of the CUL3-KEAP1 complex under hypoxia promoted PHD2 stabilization and led to significantly reduced abundance of the PHD2 target, hypoxia-inducible factor 1A (HIF1A). Thus, CUL3-KEAP1 is an essential pathway that regulates PHD2 ubiquitination and degradation in cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human KEAP1
Sigma-Aldrich
MISSION® esiRNA, targeting human CUL3
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture