Merck
  • Zika Virus Targets Glioblastoma Stem Cells through a SOX2-Integrin αvβ5 Axis.
Zika Virus Targets Glioblastoma Stem Cells through a SOX2-Integrin αvβ5 Axis.
Cell stem cell (2020-01-21)
Zhe Zhu, Pinar Mesci, Jean A Bernatchez, Ryan C Gimple, Xiuxing Wang, Simon T Schafer, Hiromi I Wettersten, Sungjun Beck, Alex E Clark, Qiulian Wu, Briana C Prager, Leo J Y Kim, Rekha Dhanwani, Sonia Sharma, Alexandra Garancher, Sara M Weis, Stephen C Mack, Priscilla D Negraes, Cleber A Trujillo, Luiz O Penalva, Jing Feng, Zhou Lan, Rong Zhang, Alex W Wessel, Sanjay Dhawan, Michael S Diamond, Clark C Chen, Robert J Wechsler-Reya, Fred H Gage, Hongzhen Hu, Jair L Siqueira-Neto, Alysson R Muotri, David A Cheresh, Jeremy N Rich
ABSTRACT
Zika virus (ZIKV) causes microcephaly by killing neural precursor cells (NPCs) and other brain cells. ZIKV also displays therapeutic oncolytic activity against glioblastoma (GBM) stem cells (GSCs). Here we demonstrate that ZIKV preferentially infected and killed GSCs and stem-like cells in medulloblastoma and ependymoma in a SOX2-dependent manner. Targeting SOX2 severely attenuated ZIKV infection, in contrast to AXL. As mechanisms of SOX2-mediated ZIKV infection, we identified inverse expression of antiviral interferon response genes (ISGs) and positive correlation with integrin αv (ITGAV). ZIKV infection was disrupted by genetic targeting of ITGAV or its binding partner ITGB5 and by an antibody specific for integrin αvβ5. ZIKV selectively eliminated GSCs from species-matched human mature cerebral organoids and GBM surgical specimens, which was reversed by integrin αvβ5 inhibition. Collectively, our studies identify integrin αvβ5 as a functional cancer stem cell marker essential for GBM maintenance and ZIKV infection, providing potential brain tumor therapy.
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, azide free, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
MISSION® Lentiviral Packaging Mix, Production of replication incompetent viral particles
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse