Merck
  • Nerve Injury-Induced Neuronal PAP-I Maintains Neuropathic Pain by Activating Spinal Microglia.
Nerve Injury-Induced Neuronal PAP-I Maintains Neuropathic Pain by Activating Spinal Microglia.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2019-11-21)
Jiayin Li, Haixiang Shi, Hui Liu, Fei Dong, Zhiping Liu, Yingjin Lu, Luonan Chen, Lan Bao, Xu Zhang
ABSTRACT
Pancreatitis-associated proteins (PAPs) display multiple functions in visceral diseases. Previous studies showed that the expression level of PAP-I was low in the DRG of naive rats but was de novo expressed after peripheral nerve injury. However, its role in neuropathic pain remains unknown. We found that PAP-I expression was continuously upregulated in the DRG neurons from rat spared nerve injury models, and transported toward the spinal dorsal horn to act as a proinflammatory factor. Intrathecal delivery of PAP-I enhanced sensory hyperalgesia, whereas PAP-I deficiency by either gene knockout or antibody application alleviated tactile allodynia at the maintenance phase after spared nerve injury. Furthermore, PAP-I functioned by activating the spinal microglia via C-C chemokine receptor Type 2 that participated in neuropathic pain. Inhibition of either microglial activation or C-C chemokine receptor Type 2 abolished the PAP-I-induced hyperalgesia. Thus, PAP-I mediates the neuron-microglial crosstalk after peripheral nerve injury and contributes to the maintenance of neuropathic pain.SIGNIFICANCE STATEMENT Neuropathic pain is maladaptive pain condition, and the maintaining mechanism is largely unclear. Here we reveal that, after peripheral nerve injury, PAP-I can be transported to the spinal dorsal horn and is crucial in the progression of neuropathic pain. Importantly, we prove that PAP-I mainly functions through activating the spinal microglia via the CCR2-p38 MAPK pathway. Furthermore, we confirm that the proinflammatory effect of PAP-I is more prominent after the establishment of neuropathic pain, thus indicating that microglia also participate in the maintenance phase of neuropathic pain.
Sigma-Aldrich
Anti-P2X3 Receptor Antibody, pain, serum, Chemicon®
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-Neurofilament 160/200 antibody, Mouse monoclonal, ~2 mg/mL, clone RMdO20, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Peripherin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-Integrin αM [CD11b] Antibody, clone OX-42, clone OX-42, Chemicon®, from mouse