Merck
  • Autophagy of an Amyloid-like Translational Repressor Regulates Meiotic Exit.
Autophagy of an Amyloid-like Translational Repressor Regulates Meiotic Exit.
Developmental cell (2020-01-29)
Fei Wang, Rudian Zhang, Wenzhi Feng, Dai Tsuchiya, Olivia Ballew, Jiajia Li, Vladimir Denic, Soni Lacefield
ABSTRACT
We explored the potential for autophagy to regulate budding yeast meiosis. Following pre-meiotic DNA replication, we blocked autophagy by chemical inhibition of Atg1 kinase or engineered degradation of Atg14 and observed homologous chromosome segregation followed by sister chromatid separation; cells then underwent additional rounds of spindle formation and disassembly without DNA re-replication, leading to aberrant chromosome segregation. Analysis of cell-cycle regulators revealed that autophagy inhibition prevents meiosis II-specific expression of Clb3 and leads to the aberrant persistence of Clb1 and Cdc5, two substrates of a meiotic ubiquitin ligase activated by Ama1. Lastly, we found that during meiosis II, autophagy degrades Rim4, an amyloid-like translational repressor whose timed clearance regulates protein production from its mRNA targets, which include CLB3 and AMA1. Strikingly, engineered Clb3 or Ama1 production restored meiotic termination in the absence of autophagy. Thus, autophagy destroys a master regulator of meiotic gene expression to enable irreversible meiotic exit.
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Prostaglandin B2, ≥98%, synthetic
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Copper(II) sulfate, ReagentPlus®, ≥99%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
JSH-23, ≥98% (HPLC), solid
Sigma-Aldrich
Bay 11-7085, ≥98% (HPLC), solid
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Supelco
Chlorpropamide, analytical standard, ≥97%
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Adenine hemisulfate salt, ≥99%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)