Merck
  • A Study into the ADP-Ribosylome of IFN-γ-Stimulated THP-1 Human Macrophage-like Cells Identifies ARTD8/PARP14 and ARTD9/PARP9 ADP-Ribosylation.
A Study into the ADP-Ribosylome of IFN-γ-Stimulated THP-1 Human Macrophage-like Cells Identifies ARTD8/PARP14 and ARTD9/PARP9 ADP-Ribosylation.
Journal of proteome research (2019-03-09)
Hideyuki Higashi, Takashi Maejima, Lang Ho Lee, Yukiyoshi Yamazaki, Michael O Hottiger, Sasha A Singh, Masanori Aikawa
ABSTRACT
ADP-ribosylation is a post-translational modification that, until recently, has remained elusive to study at the cellular level. Previously dependent on radioactive tracers to identify ADP-ribosylation targets, several advances in mass spectrometric workflows now permit global identification of ADP-ribosylated substrates. In this study, we capitalized on two ADP-ribosylation enrichment strategies, and multiple activation methods performed on the Orbitrap Fusion Lumos, to identify IFN-γ-induced ADP-ribosylation substrates in macrophages. The ADP-ribosyl binding protein, Af1521, was used to enrich ADP-ribosylated peptides, and the antipoly-ADP-ribosyl antibody, 10H, was used to enrich ADP-ribosylated proteins. ADP-ribosyl-specific mass spectra were further enriched by an ADP-ribose product ion triggered EThcD and HCD activation strategy, in combination with multiple acquisitions that segmented the survey scan into smaller ranges. HCD and EThcD resulted in overlapping and unique ADP-ribosyl peptide identifications, with HCD providing more peptide identifications but EThcD providing more reliable ADP-ribosyl acceptor sites. Our acquisition strategies also resulted in the first ever characterization of ADP-ribosyl on three poly-ADP-ribose polymerases, ARTD9/PARP9, ARTD10/PARP10, and ARTD8/PARP14. IFN-γ increased the ADP-ribosylation status of ARTD9/PARP9, ARTD8/PARP14, and proteins involved in RNA processes. This study therefore summarizes specific molecular pathways at the intersection of IFN-γ and ADP-ribosylation signaling pathways.
Sigma-Aldrich
PARP Inhibitor VIII, PJ34, The PARP Inhibitor VIII, PJ34, also referenced under CAS 344458-15-7, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.
Sigma-Aldrich
Anti-pan-ADP-ribose binding reagent, from Escherichia coli
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution