Merck
  • Effects of Klf4 and c-Myc Knockdown on Pluripotency Maintenance in Porcine Induced Pluripotent Stem Cell.
Effects of Klf4 and c-Myc Knockdown on Pluripotency Maintenance in Porcine Induced Pluripotent Stem Cell.
Cell journal (2017-11-07)
Yu-Jing Liao, Yi-Shiou Chen, Ja-Xin Lee, Lih-Ren Chen, Jenn-Rong Yang
ABSTRACT
The importance of Oct4 and Sox2 in maintaining pluripotency and self-renewal is well-understood, but the functions of Klf4 and c-Myc has not been fully investigated. In the present study, we attempted to determine the roles of Klf4 and c-Myc on pluripotency maintenance of porcine induced pluripotent stem (piPS) cells. In this experimental study, we performed short hairpin RNA (shRNA) to knock down the Klf4 and c-Myc functions of piPS cells and examined pluripotency markers and teratoma formation to evaluate piPS cell pluripotency. The shRNA-Klf4 and shRNA-c-Myc vectors containing a reporter gene, TagFP635, were transfected into piPS cells by lentivirus infection. The piPS cells fully expressing infrared fluorescence were selected to confirm gene knockdown of Klf4 and c-Myc reverse transcription-polymerase chain reaction (RT-PCR). Next, for pluripotency evaluation, expression of pluripotency markers was detected by immunocytochemical staining, and capability of teratoma formation was investigated by piPS cell transplantation into nonobese diabetic-severe combined immunodeficiency (NOD-SCID) mice. Our findings indicated that Klf4 and c-Myc functions of piPS cells were knocked down by shRNA transfection, and knockdown of Klf4 and c-Myc functions impaired expression of pluripotency markers such as Oct4, AP, SSEA-3, SSEA-4, TRA-1-6, and TRA-1-81. Furthermore, piPS cells without Klf4 and c-Myc expression failed to form teratomas. The pluripotency of piPS cells are crucially dependent upon Klf4 and c-Myc expression. These findings, suggesting potential mechanisms of Klf4 and c-Myc contribution to piPS cell formation, have important implications for application, regulation, and tumorigenesis of piPS cells.
Sigma-Aldrich
Anti-TRA-1-81 Antibody, clone TRA-1-81, clone TRA-1-81, Chemicon®, from mouse
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-4 Antibody, clone MC-813-70, clone MC-813-70, Chemicon®, from mouse
Sigma-Aldrich
Anti-Oct-4 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-TRA-2-49 Antibody, Liver/Bone/Kidney Alkaline Phosphatase, clone TRA-2-49/6E, clone TRA-2-49/6E, Chemicon®, from mouse
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse