Merck
  • Structural basis for interorganelle phospholipid transport mediated by VAT-1.
Structural basis for interorganelle phospholipid transport mediated by VAT-1.
The Journal of biological chemistry (2020-02-02)
Yasunori Watanabe, Yasushi Tamura, Chika Kakuta, Seiya Watanabe, Toshiya Endo
ABSTRACT
Eukaryotic cells are compartmentalized to form organelles, whose functions rely on proper phospholipid and protein transport. Here we determined the crystal structure of human VAT-1, a cytosolic soluble protein that was suggested to transfer phosphatidylserine, at 2.2 Å resolution. We found that VAT-1 transferred not only phosphatidylserine but also other acidic phospholipids between membranes in vitro Structure-based mutational analyses showed the presence of a possible lipid-binding cavity at the interface between the two subdomains, and two tyrosine residues in the flexible loops facilitated phospholipid transfer, likely by functioning as a gate to this lipid-binding cavity. We also found that a basic and hydrophobic loop with two tryptophan residues protruded from the molecule and facilitated binding to the acidic-lipid membranes, thereby achieving efficient phospholipid transfer.
Product Description
Avanti
18:1 NBD PE, Avanti Research - A Croda Brand
Avanti
18:1 NBD PE, Avanti Research - A Croda Brand
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1-12:0 NBD PS, Avanti Research - A Croda Brand 810195C
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand