Merck
  • Ca2+-dependent demethylation of phosphatase PP2Ac promotes glucose deprivation-induced cell death independently of inhibiting glycolysis.
Ca2+-dependent demethylation of phosphatase PP2Ac promotes glucose deprivation-induced cell death independently of inhibiting glycolysis.
Science signaling (2018-01-11)
Ha Yin Lee, Yoko Itahana, Stefan Schuechner, Masahiro Fukuda, H Shawn Je, Egon Ogris, David M Virshup, Koji Itahana
ABSTRACT
Cancer cells increase glucose metabolism to support aerobic glycolysis. However, only some cancer cells are acutely sensitive to glucose withdrawal, and the underlying mechanism of this selective sensitivity is unclear. We showed that glucose deprivation initiates a cell death pathway in cancer cells that is dependent on the kinase RIPK1. Glucose withdrawal triggered rapid plasma membrane depolarization and an influx of extracellular calcium into the cell through the L-type calcium channel Cav1.3 (CACNA1D), followed by activation of the kinase CAMK1. CAMK1 and the demethylase PPME1 were required for the subsequent demethylation and inactivation of the catalytic subunit of the phosphatase PP2A (PP2Ac) and the phosphorylation of RIPK1. Plasma membrane depolarization, PP2Ac demethylation, and cell death were prevented by glucose and, unexpectedly, by its nonmetabolizable analog 2-deoxy-d-glucose (2-DG), a glycolytic inhibitor. These findings reveal a previously unknown function of glucose as a signaling molecule that protects cells from death induced by plasma membrane depolarization, independently of its role in glycolysis. Components of this cancer cell death pathway represent potential therapeutic targets against cancer.
Sigma-Aldrich
Anti-PP2A Antibody, C subunit, Upstate®, from rabbit
Sigma-Aldrich
Anti-PP2A-methylesterase/PME-1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®