Merck
  • Alterations in histamine responses between juvenile and adult urinary bladder urothelium, lamina propria and detrusor tissues.
Alterations in histamine responses between juvenile and adult urinary bladder urothelium, lamina propria and detrusor tissues.
Scientific reports (2020-03-07)
Zane Stromberga, Russ Chess-Williams, Christian Moro
ABSTRACT
Inflammatory mediators may have a role in various lower urinary tract disorders. Histamine is known to induce significant increases in both the tension and frequency of spontaneous phasic contractions in both urothelium with lamina propria (U&LP) and detrusor muscle via the activation of H1 receptor in juvenile animal models. However, it is unclear whether age affects these contractile responses to histamine. This study assessed the histamine receptor subtypes mediating contraction in juvenile and adult porcine bladders and compared the urothelium with lamina propria and detrusor responses to histamine. Isolated tissue bath studies were conducted using strips of porcine U&LP and detrusor obtained from juvenile (6 months) and adult (3 years) animals exposed to histamine receptor agonists and antagonists. Treatment with histamine (100 µM) in U&LP of juvenile animals caused increases in baseline tension by 47.84 ± 6.52 mN/g (p < 0.001, n = 51) and by 50.76 ± 4.10 mN/g (p < 0.001, n = 55) in adult animals. Furthermore, the frequency of spontaneous phasic contractions was significantly enhanced in response to histamine in U&LP of both juvenile and adult tissues (p < 0.001 for both age groups). Treatment with an H2 agonist in U&LP of juvenile animals decreased baseline tension by 13.97 ± 3.45 mN/g (n = 12, p < 0.05), but had no effect in adult animals. Inhibition of H1 receptors resulted in significantly reduced contractile responses of U&LP and detrusor to histamine in both juvenile and adult animals (p < 0.05). Treatment with an H2 receptor antagonist significantly enhanced contractions in juvenile preparations (n = 10, p < 0.05) but had no effect in adult preparations (n = 8). In detrusor, treatment with histamine (100 µM) in juvenile tissues showed a significantly higher increase in baseline tension of 19.10 ± 4.92 mN/g (n = 51) when compared to adult tissues exhibiting increases of 8.21 ± 0.89 mN/g (n = 56, p < 0.05). The increases in the baseline tension were significantly inhibited by the presence of H1 receptor antagonists in both juvenile and adult detrusor preparations. Treatment with either the H2 receptor antagonist or agonist in detrusor had no effect on both juvenile and adult tissues. Therefore, the histamine receptor system may play an essential role in the maintenance of bladder function or in bladder dysfunction observed in some lower urinary tract disorders.
Sigma-Aldrich
Cimetidine
Sigma-Aldrich
Thioperamide maleate salt, solid
Sigma-Aldrich
Histamine dihydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Pyrilamine maleate salt