Merck
  • SUMO-1 regulates body weight and adipogenesis via PPARγ in male and female mice.
SUMO-1 regulates body weight and adipogenesis via PPARγ in male and female mice.
Endocrinology (2012-12-29)
Laura Mikkonen, Johanna Hirvonen, Olli A Jänne
ABSTRACT
Properly functioning adipose tissue is essential for normal insulin sensitivity of the body. When mice are kept on high-fat diet (HFD), adipose tissue expands, adipocytes increase in size and number, and the mice become obese. Many of these changes are mediated by the nuclear receptor peroxisome proliferator-activated receptor γ (PPARγ), the activity of which is regulated by multiple posttranslational modifications, including SUMOylation. To address the role of small ubiquitin-like modifier-1 (SUMO-1) in PPARγ function in vivo, particularly in fat cell biology, we subjected Sumo1-knockout mice to HFD. Sumo1-null mice gained less weight and had smaller and fewer adipocytes in their gonadal fat tissue on HFD, but their glucose tolerance was similar to that of wild-type littermates. Adipogenesis was impaired in Sumo1-null cells, and expression of PPARγ target genes was attenuated. In addition, both Sumo1-null cells and Sumo1-null mice responded less efficiently to rosiglitazone, a PPARγ agonist. These findings indicate that SUMO-1 is important also for transcriptional activation by the PPARγ signaling pathway and not only for trans-repressive functions of PPARγ as previously reported.
Sigma-Aldrich
ChIPAb+ N-CoR Antibody, from rabbit, purified by affinity chromatography