Merck
  • The Role of Peroxisome Proliferator–Activated Receptor Gamma (PPARγ) in Mono(2-ethylhexyl) Phthalate (MEHP)-Mediated Cytotrophoblast Differentiation.
The Role of Peroxisome Proliferator–Activated Receptor Gamma (PPARγ) in Mono(2-ethylhexyl) Phthalate (MEHP)-Mediated Cytotrophoblast Differentiation.
Environmental health perspectives (2019-02-28)
Hussein Shoaito, Julia Petit, Audrey Chissey, Nicolas Auzeil, Jean Guibourdenche, Sophie Gil, Olivier Laprévote, Thierry Fournier, Séverine A Degrelle
ABSTRACT
Phthalates are environmental contaminants commonly used as plasticizers in polyvinyl chloride (PVC) products. Recently, exposure to phthalates has been associated with preterm birth, low birth weight, and pregnancy loss. There is limited information about the possible mechanisms linking maternal phthalate exposure and placental development, but one such mechanism may be mediated by peroxisome proliferator–activated receptor γ (PPARγ). PPARγ belongs to the nuclear receptor superfamily that regulates, in a ligand-dependent manner, the transcription of target genes. Studies of PPARγ-deficient mice have demonstrated its essential role in lipid metabolism and placental development. In the human placenta, PPARγ is expressed in the villous cytotrophoblast (VCT) and is activated during its differentiation into syncytiotrophoblast. The goal of this study was to investigate the action of mono(2-ethylhexyl) phthalate (MEHP) on PPARγ activity during in vitro differentiation of VCTs. We combined immunofluorescence, PPARγ activity/hCG assays, western blotting, and lipidomics analyses to characterize the impacts of physiologically relevant concentrations of MEHP (0.1, 1, and 10 μM) on cultured VCTs isolated from human term placentas. Doses of 0.1 and 1 μM MEHP showed significantly lower PPARγ activity and less VCT differentiation in comparison with controls, whereas, surprisingly, a 10 μM dose had the opposite effect. MEHP exposure inhibited hCG production and significantly altered lipid composition. In addition, MEHP had significant effects on the mitogen-activated protein kinase (MAPK) pathway. This study suggests that MEHP has a U-shaped dose–response effect on trophoblast differentiation that is mediated by the PPARγ pathway and acts as an endocrine disruptor in the human placenta. https://doi.org/10.1289/EHP3730.
Sigma-Aldrich
Anti-Keratin 7 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Protease Inhibitor Cocktail Set I, A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
Millipore
Phosphatase Inhibitor Cocktail Set V, 50X, Phosphatase Inhibitor Cocktail Set V is a 50X cocktail of four serine/threonine and tyrosine phosphatase inhibitors.