Merck
  • Pharmacologic Blockade of αvβ1 Integrin Ameliorates Renal Failure and Fibrosis In Vivo.
Pharmacologic Blockade of αvβ1 Integrin Ameliorates Renal Failure and Fibrosis In Vivo.
Journal of the American Society of Nephrology : JASN (2017-02-22)
Yongen Chang, Wei Ling Lau, Hyunil Jo, Kazuyuki Tsujino, Leslie Gewin, Nilgun Isik Reed, Amha Atakilit, Ane Claudia Fernandes Nunes, William F DeGrado, Dean Sheppard
ABSTRACT
Activated fibroblasts are deemed the main executors of organ fibrosis. However, regulation of the pathologic functions of these cells in vivo is poorly understood. PDGF receptor β (PDGFRβ) is highly expressed in activated pericytes, a main source of fibroblasts. Studies using a PDGFRβ promoter-driven Cre system to delete αv integrins in activated fibroblasts identified these integrins as core regulators of fibroblast activity across solid organs, including the kidneys. Here, we used the same PDGFRβ-Cre line to isolate and study renal fibroblasts ex vivo We found that renal fibroblasts express three αv integrins, namely αvβ1, αvβ3, and αvβ5. Blockade of αvβ1 prevented direct binding of fibroblasts to the latency-associated peptide of TGF-β1 and prevented activation of the latent TGF-β complex. Continuous administration of a recently described potent small molecule inhibitor of αvβ1, compound 8, starting the day of unilateral ureteral obstruction operation, inhibited collagen deposition in the kidneys of mice 14 days later. Compound 8 also effectively attenuated renal failure, as measured by BUN levels in mice fed an adenine diet known to cause renal injury followed by fibrosis. Inhibition of αvβ1 integrin could thus hold promise as a therapeutic intervention in CKD characterized by renal fibrosis.
Sigma-Aldrich
Anti-mouse Integrin alphaV Antibody, clone RMV-7 (Preservative free), clone RMV-7, from rat
Sigma-Aldrich
Anti-Integrin beta1 Antibody, Cytosolic, serum, Chemicon®