Merck
  • LRP1 integrates murine macrophage cholesterol homeostasis and inflammatory responses in atherosclerosis.
LRP1 integrates murine macrophage cholesterol homeostasis and inflammatory responses in atherosclerosis.
eLife (2017-11-17)
Xunde Xian, Yinyuan Ding, Marco Dieckmann, Li Zhou, Florian Plattner, Mingxia Liu, John S Parks, Robert E Hammer, Philippe Boucher, Shirling Tsai, Joachim Herz
ABSTRACT
Low-density lipoprotein receptor-related protein 1 (LRP1) is a multifunctional cell surface receptor with diverse physiological roles, ranging from cellular uptake of lipoproteins and other cargo by endocytosis to sensor of the extracellular environment and integrator of a wide range of signaling mechanisms. As a chylomicron remnant receptor, LRP1 controls systemic lipid metabolism in concert with the LDL receptor in the liver, whereas in smooth muscle cells (SMC) LRP1 functions as a co-receptor for TGFβ and PDGFRβ in reverse cholesterol transport and the maintenance of vascular wall integrity. Here we used a knockin mouse model to uncover a novel atheroprotective role for LRP1 in macrophages where tyrosine phosphorylation of an NPxY motif in its intracellular domain initiates a signaling cascade along an LRP1/SHC1/PI3K/AKT/PPARγ/LXR axis to regulate and integrate cellular cholesterol homeostasis through the expression of the major cholesterol exporter ABCA1 with apoptotic cell removal and inflammatory responses.
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
Anti-Apolipoprotein E Goat pAb, liquid, Calbiochem®
Sigma-Aldrich
PDGF-BB, recombinant human, The Platelet Derived Growth Factor (PDGF) is a potent mitogen for a wide range of cell types including fibroblasts, smooth muscle & connective tissue.
Sigma-Aldrich
Anti-Apolipoprotein B Goat pAb, liquid, Calbiochem®
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests
Sigma-Aldrich
Akt1/2 kinase inhibitor, ≥98% (HPLC)
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)