Merck
Slow activation of fast mitochondrial Ca2+ uptake by cytosolic Ca2.
The Journal of biological chemistry (2018-09-20)
Emy Basso, Giulia Rigotto, Andrés E Zucchetti, Tullio Pozzan
ABSTRACT
Mitochondrial Ca2+ uptake through the mitochondrial Ca2+ uniporter (MCU) is a tightly controlled process that sustains cell functions mainly by fine-tuning oxidative metabolism to cellular needs. The kinetics of Ca2+ fluxes across the mitochondrial membranes have been studied both in vitro and in vivo for many years, and the discovery of the molecular components of the MCU has further clarified that this Ca2+ uptake mechanism is based on a complex system subject to elaborate layers of controls. Alterations in the speed or capacity of the in-and-out pathways can have detrimental consequences for both the organelle and the cell, impairing cellular metabolism and ultimately causing cell death. Here, we report that pretreatment of deenergized mitochondria with low-micromolar Ca2+ concentrations for a few minutes markedly increases the speed of mitochondrial Ca2+ uptake upon re-addition of an oxidizable substrate. We found that this phenomenon is sensitive to alterations in the level of the MCU modulator proteins mitochondrial calcium uptake 1 (MICU1) and 2 (MICU2), and is accompanied by changes in the association of MICU1-MICU2 complexes with MCU. This increased Ca2+ uptake capacity, occurring under conditions mimicking those during ischemia/reperfusion in vivo, could lead to a massive amount of Ca2+ entering the mitochondrial matrix even at relatively low levels of cytosolic Ca2+ We conclude that the phenomenon uncovered here represents a potential threat of mitochondrial Ca2+ overload to the cell.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-IMPA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution