Merck
  • Dickkopf Wnt signaling pathway inhibitor 1 regulates the differentiation of mouse embryonic stem cells in vitro and in vivo.
Dickkopf Wnt signaling pathway inhibitor 1 regulates the differentiation of mouse embryonic stem cells in vitro and in vivo.
Molecular medicine reports (2015-12-10)
Liping Ou, Liaoqiong Fang, Hejing Tang, Hai Qiao, Xiaomei Zhang, Zhibiao Wang
ABSTRACT
Embryonic stem cells (ESCs) are pluripotent stem cells derived from early stage embryos. It remains unclear whether inhibiting the Wnt/β‑catenin signaling pathway using dickkopf Wnt signaling pathway inhibitor 1 (DKK1) impacts on the differentiation potential of mouse ESCs in vitro and in vivo. In the present study, immunohistochemical staining was used to measure the expression of markers of the three germ layers in ESCs and teratomas derived from ESCs. The expression of markers for the Wnt/β‑catenin signaling pathway were detected by reverse transcription‑polymerase chain reaction (RT‑qPCR). Immunohistochemistry and western blotting indicated that the expression levels of octamer‑binding transcription factor 4 in the DKK1‑treated ESC group were significantly greater compared with the control ESCs. Reduced expression levels of NeuroD and bone morphogenetic protein 4 were observed in the DKK1‑treated ESCs and teratomas derived from DKK1‑treated ESCs compared with the control group. Increased expression levels of SOX17 were observed in the DKK1‑treated ESCs compared with the control group. RT‑qPCR indicated that β‑catenin expression was significantly reduced in DKK1‑treated ESCs and teratomas derived from DKK1‑treated ESCs compared with the control groups. Western blotting indicated no alterations in the expression of GSK‑3β, however, the levels of phosphorylated‑GSK‑3β were significantly greater in the DKK1 treatment groups, while cyclin D1 and c‑Myc expression levels were significantly reduced in the DKK1 treatment groups compared with the control groups. These results suggest that inhibiting Wnt signaling in ESCs using DKK1 may promote mouse ESCs to differentiate into endoderm in vitro and in vivo, and suppress the tumorigenicity of ESCs.
Sigma-Aldrich
Anti-Cyclin D Antibody, from rabbit, purified by affinity chromatography