Merck
  • Roflumilast Reduces Cerebral Inflammation in a Rat Model of Experimental Subarachnoid Hemorrhage.
Roflumilast Reduces Cerebral Inflammation in a Rat Model of Experimental Subarachnoid Hemorrhage.
Inflammation (2017-04-30)
Qingjian Wu, Lifeng Qi, Hanxia Li, Leilei Mao, Mingfeng Yang, Rongxia Xie, Xiaoyi Yang, Jian Wang, Zongyong Zhang, Jiming Kong, Baoliang Sun
ABSTRACT
Roflumilast, a selective inhibitor for PDE4, is approved by FDA as an anti-inflammation drug for treatment of chronic obstructive pulmonary disease (COPD). This study investigates the effects of roflumilast on cerebral inflammation in the rat SAH model. Here, we show that subcutaneous administration of roflumilast (3 mg/kg) significantly improved the neurological deficits. Measurement of evans blue extravasation and brain water content revealed a significant reduction of blood-brain barrier permeability and brain edema. Importantly, roflumilast treatment remarkably decreased levels of IL-1β, IL-6, and TNF-α and the number of apoptotic neurons in the brain after SAH. These results indicate that roflumilast is effective in treating cerebral inflammation following SAH.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−TRITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–TRITC antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Sigma-Aldrich
Anti-NeuN (rabbit monoclonal) Antibody, clone 27-4, clone 27-4, from rabbit