Merck
Perinatal changes in mitral and aortic valve structure and composition.
Pediatric and developmental pathology : the official journal of the Society for Pediatric Pathology and the Paediatric Pathology Society (2010-06-12)
Elizabeth H Stephens, Allison D Post, Daniel R Laucirica, K Jane Grande-Allen
ABSTRACT
At birth, the mechanical environment of valves changes radically as fetal shunts close and pulmonary and systemic vascular resistances change. Given that valves are reported to be mechanosensitive, we investigated remodeling induced by perinatal changes by examining compositional and structural differences of aortic and mitral valves (AVs, MVs) between 2-day-old and 3rd fetal trimester porcine valves using immunohistochemistry and Movat pentachrome staining. Aortic valve composition changed more with birth than the MV, consistent with a greater change in AV hemodynamics. At 2 days, AV demonstrated a trend of greater versican and elastin (P  =  0.055), as well as greater hyaluronan turnover (hyaluronan receptor for endocytosis, P  =  0.049) compared with the 3rd-trimester samples. The AVs also demonstrated decreases in proteins related to collagen synthesis and fibrillogenesis with birth, including procollagen I, prolyl 4-hydroxylase, biglycan (all P ≤ 0.005), and decorin (P  =  0.059, trend). Both AVs and MVs demonstrated greater delineation between the leaflet layers in 2-day-old compared with 3rd-trimester samples, and AVs demonstrated greater saffron-staining collagen intensity, suggesting more mature collagen in 2-day-old compared with 3rd-trimester samples (each P < 0.05). The proportion of saffron-staining collagen also increased in AV with birth (P < 0.05). The compositional and structural changes that occur with birth, as noted in this study, likely are important to proper neonatal valve function. Furthermore, normal perinatal changes in hemodynamics often do not occur in congenital valve disease; the corresponding perinatal matrix maturation may also be lacking and could contribute to poor function of congenitally malformed valves.
Sigma-Aldrich
Anti-MMP-13 Antibody, clone 181-15A12, clone 181-15A12, Chemicon®, from mouse