Merck
  • ZFAND1 Recruits p97 and the 26S Proteasome to Promote the Clearance of Arsenite-Induced Stress Granules.
ZFAND1 Recruits p97 and the 26S Proteasome to Promote the Clearance of Arsenite-Induced Stress Granules.
Molecular cell (2018-05-29)
Ankit Turakhiya, Susanne R Meyer, Gabriella Marincola, Stefanie Böhm, Jens T Vanselow, Andreas Schlosser, Kay Hofmann, Alexander Buchberger
ABSTRACT
Stress granules (SGs) are cytoplasmic assemblies of mRNPs stalled in translation initiation. They are induced by various stress conditions, including exposure to the environmental toxin and carcinogen arsenic. While perturbed SG turnover is linked to the pathogenesis of neurodegenerative diseases, the molecular mechanisms underlying SG formation and turnover are still poorly understood. Here, we show that ZFAND1 is an evolutionarily conserved regulator of SG clearance. ZFAND1 interacts with two key factors of protein degradation, the 26S proteasome and the ubiquitin-selective segregase p97, and recruits them to arsenite-induced SGs. In the absence of ZFAND1, SGs lack the 26S proteasome and p97, accumulate defective ribosomal products, and persist after arsenite removal, indicating their transformation into aberrant, disease-linked SGs. Accordingly, ZFAND1 depletion is epistatic to the expression of pathogenic mutant p97 with respect to SG clearance, suggesting that ZFAND1 function is relevant to the multisystem degenerative disorder IBMPFD/ALS.
Sigma-Aldrich
Anti-ZFAND1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution