Merck
  • Manipulating mtDNA in vivo reprograms metabolism via novel response mechanisms.
Manipulating mtDNA in vivo reprograms metabolism via novel response mechanisms.
PLoS genetics (2019-10-05)
Diana Bahhir, Cagri Yalgin, Liina Ots, Sampsa Järvinen, Jack George, Alba Naudí, Tatjana Krama, Indrikis Krams, Mairi Tamm, Ana Andjelković, Eric Dufour, Jose M González de Cózar, Mike Gerards, Mikael Parhiala, Reinald Pamplona, Howard T Jacobs, Priit Jõers
ABSTRACT
Mitochondria have been increasingly recognized as a central regulatory nexus for multiple metabolic pathways, in addition to ATP production via oxidative phosphorylation (OXPHOS). Here we show that inducing mitochondrial DNA (mtDNA) stress in Drosophila using a mitochondrially-targeted Type I restriction endonuclease (mtEcoBI) results in unexpected metabolic reprogramming in adult flies, distinct from effects on OXPHOS. Carbohydrate utilization was repressed, with catabolism shifted towards lipid oxidation, accompanied by elevated serine synthesis. Cleavage and translocation, the two modes of mtEcoBI action, repressed carbohydrate rmetabolism via two different mechanisms. DNA cleavage activity induced a type II diabetes-like phenotype involving deactivation of Akt kinase and inhibition of pyruvate dehydrogenase, whilst translocation decreased post-translational protein acetylation by cytonuclear depletion of acetyl-CoA (AcCoA). The associated decrease in the concentrations of ketogenic amino acids also produced downstream effects on physiology and behavior, attributable to decreased neurotransmitter levels. We thus provide evidence for novel signaling pathways connecting mtDNA to metabolism, distinct from its role in supporting OXPHOS.
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Digitonin, ~50% (TLC)
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Methyl trifluoroacetate, 99%
Sigma-Aldrich
Triglyceride Reagent, used for quantitative enzymatic determination of triglyerides
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Supelco
Glucose Assay Reagent, for use with enzymatic assay kits GAHK20, SCA20, FA20, SA20
Sigma-Aldrich
Trehalase from porcine kidney, buffered aqueous glycerol solution, ≥1.0 units/mg protein
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Deepwell 96 well plate, polypropylene, sterile, pack of 24 ea