Merck
  • Photoacoustic simulations of microvascular bleeding: spectral analysis and its application for monitoring vascular-targeted treatments.
Photoacoustic simulations of microvascular bleeding: spectral analysis and its application for monitoring vascular-targeted treatments.
Journal of biomedical optics (2019-11-11)
Muhannad N Fadhel, Eno Hysi, Jason Zalev, Michael C Kolios
ABSTRACT
Solid tumors are typically supplied nutrients by a network of irregular blood vessels. By targeting these vascular networks, it might be possible to hinder cancer growth and metastasis. Vascular disrupting agents induce intertumoral hemorrhaging, making photoacoustic (PA) imaging well positioned to detect bleeding due to its sensitivity to hemoglobin and its various states. We introduce a fractal-based numerical model of intertumoral hemorrhaging to simulate the PA signals from disrupted tumor blood vessels. The fractal model uses bifurcated cylinders to represent vascular trees. To mimic bleeding from blood vessels, hemoglobin diffusion from microvessels was simulated. In the simulations, the PA signals were detected by a linear array transducer (30 MHz center frequency) of four different vascular trees. The power spectrum of each beamformed PA signal was computed and fitted to a straight line within the −6-dB bandwidth of the receiving transducer. The spectral slope and midband fit (MBF) based on the fit decreased by 0.11  dB  /  MHz and 2.12 dB, respectively, 1 h post bleeding, while the y-intercept increased by 1.21 dB. The results suggest that spectral PA analysis can be used to measure changes in the concentration and spatial distribution of hemoglobin in tissue without the need to resolve individual vessels. The simulations support the feasibility of using PA imaging and spectral analysis in cancer treatment monitoring by detecting microvessel disruption.
Sigma-Aldrich
Nigrosin, certified by the BSC
Sigma-Aldrich
Intralipid, 20%, emulsion
Sigma-Aldrich
Tubocurarine hydrochloride pentahydrate, ≥97%
Sigma-Aldrich
Magnesium methyl carbonate solution, 2.0 M in DMF
Sigma-Aldrich
Cardiogreen, polymethine dye
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent, Molecular Biology
Sigma-Aldrich
N749 Black Dye