Merck
  • Selective Inhibition of Spindle Microtubules by a Tubulin-Binding Quinazoline Derivative.
Selective Inhibition of Spindle Microtubules by a Tubulin-Binding Quinazoline Derivative.
Molecular pharmacology (2019-09-01)
Jun-Ichi Sawada, Hirosuke Ishii, Kenji Matsuno, Masayuki Sato, Yumiko Suzuki, Akira Asai
ABSTRACT
In the research field of tubulin-binding agents for the development of anticancer agents, hidden targets are emerging as a problem in understanding the exact mechanisms of actions. The quinazoline derivative 1-(4-methoxyphenyl)-1-(quinazolin-4-yl)ethan-1-ol (PVHD121) has anti-cell proliferative activity and inhibits tubulin polymerization by binding to the colchicine site of tubulin. However, the molecular mechanism of action of PVHD121 in cells remains unclear. Here, we demonstrate that PVHD121 delays mitotic entry and efficiently causes mitotic arrest with spindle checkpoint activation, leading to subsequent cell death. The dominant phenotype induced by PVHD121 was aberrant spindles with robust microtubules and unseparated centrosomes. The microtubules were radially distributed, and their ends appeared to adhere to kinetochores, and not to centrosomes. Extensive inhibition by high concentrations of PVHD121 eliminated all microtubules from cells. PVHD277 [1-(4-methoxyphenyl)-1-(2-morpholinoquinazolin-4-yl)ethan-1-ol], a PVHD121 derivative with fluorescence, tended to localize close to the centrosomes when cells prepared to enter mitosis. Our results show that PVHD121 is an antimitotic agent that selectively disturbs microtubule formation at centrosomes during mitosis. This antimitotic activity can be attributed to the targeting of centrosome maturation in addition to the interference with microtubule dynamics. Due to its unique bioactivity, PVHD121 is a potential tool for studying the molecular biology of mitosis and a potential lead compound for the development of anticancer agents. SIGNIFICANCE STATEMENT: Many tubulin-binding agents have been developed as potential anticancer agents. The aim of this study was to understand the subcellular molecular actions of a quinazoline derivative tubulin-binding agent, 1-(4-methoxyphenyl)-1-(quinazolin-4-yl)ethan-1-ol (PVHD121). As expected from its binding activity to tubulin, PVHD121 caused aberrant spindles and inhibited mitotic progression. However, in addition to tubulin, PVHD121 also targeted an unexpected biomolecule involved in centrosome maturation. Due to targeting the biomolecule just before entering mitosis, PVHD121 preferentially inhibited centrosome-derived microtubules rather than chromosome-derived microtubules during spindle formation. This study not only revealed the molecular action of PVHD121 in cells but also emphasized the importance of considering possible tubulin-independent effects of tubulin-binding agents via hidden targeted biomolecules for future use.
Sigma-Aldrich
PVHD121, ≥98% (HPLC)
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, purified from hybridoma cell culture
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder